siRNA Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145205.5 | NP_660206.2 | 2 | 1822 | ||
GenBank mRNA | AK097707.1 | - | 2 | 1823 | ||
AK130859.1 | - | 1 | 241 | |||
AK311985.1 | - | 1 | 208 | |||
BC021180.2 | - | 1 | 192 | 5266418 | ||
BI827031.1 | - | 1 | 181 | |||
DQ207367.1 | - | 1 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
