siRNA Details
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083913.1 | NP_001077382.1 | 4 | 815 | ||
NM_017787.4 | NP_060257.4 | 4 | 928 | |||
XM_011539913.2 | XP_011538215.1 | 4 | 719 | |||
XM_017016360.1 | XP_016871849.1 | 4 | 2338 | |||
GenBank mRNA | AK000374.1 | - | 1 | 293 | ||
AK056285.1 | - | 4 | 812 | |||
AK300871.1 | - | 4 | 860 | |||
AK316452.1 | - | 4 | 827 | |||
AY656171.1 | - | 4 | 812 | |||
BC113857.1 | - | 4 | 792 | 40027916 | ||
BC114465.1 | - | 4 | 792 | 40027914 | ||
BQ924256.1 | - | 1 | 32 | |||
CX163170.1 | - | 4 | 761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
