siRNA Details
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244808.1 | NP_001231737.1 | 7 | 834 | ||
NM_001244810.1 | NP_001231739.1 | 7 | 731 | |||
NM_001244812.1 | NP_001231741.1 | 4 | 359 | |||
NM_001244814.1 | NP_001231743.1 | 3 | 528 | |||
NM_001244815.1 | NP_001231744.1 | 2 | 469 | |||
NM_001244816.1 | NP_001231745.1 | 6 | 636 | |||
NM_032682.5 | NP_116071.2 | 7 | 731 | |||
XM_005264735.3 | XP_005264792.1 | 2 | 420 | |||
XM_005264736.3 | XP_005264793.1 | 2 | 469 | |||
XM_005264737.4 | XP_005264794.1 | 7 | 740 | |||
XM_006713102.2 | XP_006713165.1 | 6 | 692 | |||
XM_006713103.2 | XP_006713166.1 | 4 | 429 | |||
XM_006713104.2 | XP_006713167.1 | 5 | 662 | |||
XM_011533584.2 | XP_011531886.1 | 7 | 802 | |||
XM_011533585.2 | XP_011531887.1 | 6 | 654 | |||
XM_011533588.2 | XP_011531890.1 | 2 | 252 | |||
XM_017006165.1 | XP_016861654.1 | 6 | 573 | |||
XM_017006166.1 | XP_016861655.1 | 6 | 669 | |||
XM_017006167.1 | XP_016861656.1 | 2 | 469 | |||
GenBank mRNA | AF146696.1 | - | 5 | 468 | ||
AF250920.1 | - | 6 | 636 | |||
AK027264.1 | - | 2 | 101 | |||
AK074321.1 | - | 3 | 527 | |||
AK092383.1 | - | 4 | 359 | |||
AK122710.1 | - | 7 | 834 | |||
AK297649.1 | - | 2, 3 | 114 | |||
BC131720.1 | - | 4 | 356 | |||
BX647682.1 | - | 2 | 117 | |||
DA634017.1 | - | 2 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
