siRNA Details
Gene Symbol: MUC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033049.3 | NP_149038.3 | 2 | 219 | ||
GenBank mRNA | AB035807.1 | - | 2 | 204 | ||
AF286113.1 | - | 2 | 207 | |||
AY358831.1 | - | 2 | 204 | |||
DA428072.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
