siRNA Details
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000269.1 | - | 6 | 988 | ||
AL713728.1 | - | 3 | 3866 | |||
BC047566.1 | - | 6 | 1585 | 5259068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
