siRNA Details
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 20 | 2410 | ||
NM_001204505.2 | NP_001191434.1 | 17 | 2031 | |||
NM_001204506.2 | NP_001191435.1 | 17 | 2031 | |||
NM_001297429.1 | NP_001284358.1 | 15 | 1798 | |||
NM_152910.5 | NP_690874.2 | 19 | 2373 | |||
NM_178009.4 | NP_821077.1 | 19 | 2373 | |||
NR_123714.1 | 17 | 2031 | ||||
NR_123715.1 | 20 | 2839 | ||||
GenBank mRNA | AB078967.1 | - | 19 | 2328 | ||
AB078968.1 | - | 19 | 2328 | |||
AK098302.1 | - | 4 | 392 | |||
AK122909.1 | - | 17 | 1996 | |||
AK289546.1 | - | 20 | 2412 | |||
AK302631.1 | - | 15 | 1758 | |||
AK302727.1 | - | 17 | 2031 | |||
AL833883.1 | - | 5 | 461 | |||
BC043292.2 | - | 17 | 1988 | 5297895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
