siRNA Details
Gene Symbol: GRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002086.4 | NP_002077.1 | 4 | 572 | ||
GenBank mRNA | AF498925.1 | - | 3 | 215 | ||
AK091010.1 | - | 4 | 1000 | |||
AK315565.1 | - | 4 | 568 | |||
BC000631.2 | - | 4 | 517 | 3345524 | ||
BI459414.1 | - | 4 | 579 | |||
CR450363.1 | - | 3 | 215 | |||
CR541942.1 | - | 3 | 215 | |||
M96995.1 | - | 3 | 293 | |||
X62852.1 | - | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
