siRNA Details
Gene Symbol: MUC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204287.1 | NP_001191216.1 | 2, 3 | 257 | ||
NM_001204293.1 | NP_001191222.1 | 2, 3 | 230 | |||
NM_001204297.1 | NP_001191226.1 | 2, 3 | 257 | |||
NM_002456.5 | NP_002447.4 | 2, 3 | 230 | |||
GenBank mRNA | AF348143.1 | - | 2, 3 | 182 | ||
EF583643.1 | - | 2, 3 | 334 | |||
EF583647.1 | - | 2, 3 | 235 | |||
EF670707.1 | - | 2, 3 | 199 | |||
EF670711.1 | - | 2, 3 | 208 | |||
EF670718.1 | - | 2, 3 | 235 | |||
FJ226037.1 | - | 2, 3 | 334 | |||
FJ226042.1 | - | 2, 3 | 235 | |||
FJ226046.1 | - | 2, 3 | 307 | |||
FJ226049.1 | - | 2, 3 | 235 | |||
FJ226050.1 | - | 2, 3 | 208 | |||
U60261.1 | - | 2, 3 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
