siRNA Details
Gene Symbol: GIMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015660.2 | NP_056475.1 | 3 | 162 | ||
GenBank mRNA | AK074199.1 | - | 3 | 161 | ||
AL110151.1 | - | 3 | 141 | |||
BC013934.1 | - | 3 | 162 | 3950542 | ||
BC032345.1 | - | 3 | 138 | 5208411 | ||
BG687383.1 | - | 3 | 163 |
