siRNA Details
Gene Symbol: B3GAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018644.3 | NP_061114.2 | 5 | 1075 | ||
NM_054025.2 | NP_473366.1 | 4 | 1207 | |||
XM_005271506.3 | XP_005271563.1 | 5 | 1126 | |||
XM_011542751.2 | XP_011541053.1 | 6 | 1338 | |||
XM_011542753.2 | XP_011541055.1 | 6 | 1341 | |||
XM_017017550.1 | XP_016873039.1 | 4 | 963 | |||
XM_017017551.1 | XP_016873040.1 | 5 | 2900 | |||
XM_017017552.1 | XP_016873041.1 | 3 | 1662 | |||
GenBank mRNA | AB029396.1 | - | 5 | 989 | ||
AK299637.1 | - | 4 | 893 | |||
AL834455.1 | - | 2 | 383 | |||
BC010466.1 | - | 4 | 1207 | 4157589 | ||
CR457098.1 | - | 3 | 814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
