siRNA Details
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024302.4 | NP_077278.1 | 3, 4 | 713 | ||
NM_032950.3 | NP_116568.1 | 3, 4 | 713 | |||
NR_111988.1 | 3, 4 | 713 | ||||
XM_011525226.2 | XP_011523528.1 | 4, 5 | 825 | |||
XM_011525227.1 | XP_011523529.1 | 4, 5 | 454 | |||
XM_011525228.1 | XP_011523530.1 | 4, 5 | 479 | |||
XM_011525229.2 | XP_011523531.1 | 3, 4 | 354 | |||
XM_011525230.1 | XP_011523532.1 | 3, 4 | 713 | |||
XM_011525231.1 | XP_011523533.1 | 3, 4 | 713 | |||
XM_011525232.2 | XP_011523534.2 | 3, 4 | 713 | |||
XM_017025061.1 | XP_016880550.1 | 4, 5 | 521 | |||
XM_017025062.1 | XP_016880551.1 | 3, 4 | 713 | |||
XM_017025063.1 | XP_016880552.1 | 3, 4 | 713 | |||
XM_017025064.1 | XP_016880553.1 | 3, 4 | 713 | |||
GenBank mRNA | AF219624.1 | - | 3, 4 | 552 | ||
AF315683.1 | - | 3, 4 | 398 | |||
AF330002.1 | - | 3, 4 | 658 | |||
AK122604.1 | - | 3, 4 | 713 | |||
AY358987.1 | - | 3, 4 | 588 | |||
BC002631.2 | - | 3, 4 | 630 | 3610296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
