siRNA Details
Gene Symbol: GMCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178439.4 | NP_848526.1 | 2 | 628 | ||
XM_011533033.1 | XP_011531335.1 | 2 | 592 | |||
XM_011533034.2 | XP_011531336.1 | 2 | 574 | |||
XM_017004704.1 | XP_016860193.1 | 2 | 527 | |||
GenBank mRNA | AK023119.1 | - | 2 | 534 | ||
AK024042.1 | - | 2 | 592 | |||
BC007420.2 | - | 2 | 548 | 3030159 |
Gene Symbol: GMCL1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003281.1 | 1 | 474 | |||
GenBank mRNA | AK315701.1 | - | 1 | 444 | ||
BC024184.2 | - | 1 | 460 | |||
BC024185.1 | - | 1 | 468 | |||
BC033886.2 | - | 1 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
