siRNA Details
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 6 | 804 | ||
NM_002295.5 | NP_002286.2 | 6 | 789 | |||
GenBank mRNA | AF503367.1 | - | 1 | 749 | ||
AK055991.1 | - | 2 | 1572 | |||
BC005391.1 | - | 1 | 761 | 3997019 | ||
BC008867.2 | - | 1 | 710 | 4130936 | ||
BC010418.2 | - | 1 | 741 | 4079845 | ||
BC013827.2 | - | 1 | 745 | 3446816 | ||
BC018867.2 | - | 1 | 735 | |||
BC034537.1 | - | 1 | 742 | 3455545 | ||
BC050688.1 | - | 1 | 744 | 4443949 | ||
BC053370.1 | - | 1 | 742 | 5108337 | ||
BC062714.1 | - | 1 | 765 | 3686924 | ||
BC066941.1 | - | 6 | 758 | 4820432 | ||
BC068062.1 | - | 1 | 723 | 5427114 | ||
BC070263.1 | - | 6 | 789 | 3996864 | ||
BC071693.1 | - | 1 | 751 | 3884457 | ||
BC071968.1 | - | 1 | 742 | 5814758 | ||
BC071969.1 | - | 1 | 730 | 6483856 | ||
BC071970.1 | - | 1 | 715 | 6484091 | ||
BC073863.1 | - | 1 | 727 | 3924066 | ||
BC107567.1 | - | 1 | 762 | 3681016 | ||
BT007219.1 | - | 5 | 680 | |||
J03799.1 | - | 5 | 717 | |||
M14199.1 | - | 3 | 200 | |||
X15005.1 | - | 6 | 758 | |||
X61156.1 | - | 5 | 650 |
Gene Symbol: RPSAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648992.1 | - | 2 | 2235 |
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 1103 |
Gene Symbol: RPSAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026890.1 | 1 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
