siRNA Details
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 27 | 6222 | ||
XM_005263115.3 | XP_005263172.1 | 27 | 6277 | |||
XM_011532091.2 | XP_011530393.1 | 26 | 6171 | |||
XM_011532092.2 | XP_011530394.1 | 26 | 6102 | |||
XM_011532093.2 | XP_011530395.1 | 26 | 6048 | |||
XM_011532094.2 | XP_011530396.1 | 25 | 5964 | |||
XM_011532097.2 | XP_011530399.1 | 20 | 3324 | |||
XM_011532098.2 | XP_011530400.1 | 18 | 2998 | |||
XM_011532099.2 | XP_011530401.1 | 17 | 2957 | |||
XM_017008369.1 | XP_016863858.1 | 26 | 6217 | |||
XM_017008370.1 | XP_016863859.1 | 25 | 5997 | |||
XM_017008371.1 | XP_016863860.1 | 24 | 5877 | |||
XM_017008372.1 | XP_016863861.1 | 24 | 5256 | |||
XM_017008373.1 | XP_016863862.1 | 23 | 5193 | |||
GenBank mRNA | AK090556.1 | - | 9 | 1857 | ||
AK126054.1 | - | 16 | 2606 | |||
AL137700.1 | - | 12 | 1669 | |||
BX647594.1 | - | 15 | 2577 |
