siRNA Details
Gene Symbol: CEP128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152446.3 | NP_689659.2 | 4 | 477 | ||
XM_006720056.3 | XP_006720119.1 | 5 | 658 | |||
XM_011536490.2 | XP_011534792.1 | 5 | 658 | |||
XM_011536491.2 | XP_011534793.1 | 5 | 658 | |||
XM_011536492.2 | XP_011534794.1 | 5 | 499 | |||
XM_011536493.2 | XP_011534795.1 | 5 | 658 | |||
XM_011536495.2 | XP_011534797.1 | 5 | 658 | |||
XM_017021042.1 | XP_016876531.1 | 6 | 776 | |||
GenBank mRNA | AK309833.1 | - | 5 | 601 | ||
BC045834.1 | - | 5 | 584 | |||
BC150610.1 | - | 3 | 335 | 9056980 | ||
BX248746.1 | - | 6 | 638 | |||
HQ215523.1 | - | 5 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
