siRNA Details
Gene Symbol: HS6ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153456.3 | NP_703157.2 | 2 | 6188 | ||
GenBank mRNA | AK091343.1 | - | 1 | 439 | ||
BC037795.1 | - | 1 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
