siRNA Details
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321547.1 | NP_001308476.1 | 7 | 1256 | ||
NM_001321548.1 | NP_001308477.1 | 7 | 1234 | |||
NM_001321550.1 | NP_001308479.1 | 7 | 1617 | |||
NM_152387.3 | NP_689600.2 | 7 | 1639 | |||
GenBank mRNA | AK095137.1 | - | 1 | 668 | ||
AK290867.1 | - | 7 | 1230 | |||
BC045189.1 | - | 7 | 1610 | 5296647 | ||
BC059366.1 | - | 7 | 1587 | |||
BC067755.1 | - | 7 | 1230 | 5299303 | ||
BU181789.1 | - | 3 | 495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
