siRNA Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164522.2 | NP_001157994.1 | 5 | 656 | ||
NM_177403.5 | NP_796377.3 | 4 | 730 | |||
XM_006711288.3 | XP_006711351.1 | 5 | 653 | |||
GenBank mRNA | AK127140.1 | - | 4 | 714 | ||
AY094596.1 | - | 5 | 603 | |||
BC017092.1 | - | 5 | 598 | 3851998 |
