siRNA Details
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 565 | |||
GenBank mRNA | AK292266.1 | - | 2 | 541 | ||
BC067900.1 | - | 2 | 582 | |||
BC117492.1 | - | 2 | 439 | |||
BC117494.1 | - | 2 | 438 |
Gene Symbol: LOC643406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029405.1 | 2 | 564 | |||
GenBank mRNA | AK125948.1 | - | 2 | 554 | ||
BC031676.1 | - | 2 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
