siRNA Details
Gene Symbol: C17orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038704.2 | NP_001033793.2 | 1 | 977 | ||
XM_011524637.2 | XP_011522939.1 | 1 | 977 | |||
XM_017024498.1 | XP_016879987.1 | 1 | 977 | |||
GenBank mRNA | AK097440.1 | - | 1 | 927 | ||
BC022189.2 | - | 1 | 973 | 4824904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
