siRNA Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 5 | 1639 | ||
XM_005250181.3 | XP_005250238.1 | 5 | 1713 | |||
XM_006715867.3 | XP_006715930.1 | 5 | 1718 | |||
XM_006715868.3 | XP_006715931.1 | 5 | 1713 | |||
XM_006715869.3 | XP_006715932.1 | 4 | 1622 | |||
XM_011515859.2 | XP_011514161.1 | 5 | 1434 | |||
GenBank mRNA | AF218032.1 | - | 5 | 1609 | ||
AK297946.1 | - | 3 | 827 | |||
AK309674.1 | - | 5 | 1633 | |||
BC037348.1 | - | 1 | 613 | |||
BC047641.1 | - | 5 | 1658 | |||
BC110623.1 | - | 5 | 1184 | 40036036 | ||
BC110624.1 | - | 5 | 1179 | 40036038 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081678, BC119236, BC066073, BC119262, BC144761, and others.
Gene Ontology Categories:
