siRNA Details
Gene Symbol: FASTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258461.1 | NP_001245390.1 | 2 | 545 | ||
NM_006712.4 | NP_006703.1 | 2 | 545 | |||
XM_005249932.1 | XP_005249989.1 | 2 | 737 | |||
XM_005249933.3 | XP_005249990.1 | 2 | 943 | |||
XM_011515761.2 | XP_011514063.1 | 2 | 545 | |||
XM_011515762.2 | XP_011514064.1 | 2 | 545 | |||
XM_017011704.1 | XP_016867193.1 | 2 | 737 | |||
GenBank mRNA | AB209783.1 | - | 2 | 446 | ||
AK023141.1 | - | 2 | 931 | |||
AK223132.1 | - | 2 | 455 | |||
AK292232.1 | - | 2 | 430 | |||
AL713798.1 | - | 1 | 920 | |||
BC000377.2 | - | 2 | 457 | 2820338 | ||
BC011770.2 | - | 2 | 456 | 3831196 | ||
DA253544.1 | - | 2 | 545 | |||
X86779.1 | - | 2 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
