siRNA Details
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 2, 3 | 229 | ||
NM_138635.3 | NP_619541.1 | 2, 3 | 229 | |||
NM_201516.2 | NP_958924.1 | 2, 3 | 229 | |||
GenBank mRNA | AB209001.1 | - | 2, 3 | 140 | ||
AF081192.1 | - | 2, 3 | 94 | |||
AK023973.1 | - | 2, 3 | 180 | |||
AK129989.1 | - | 2, 3 | 149 | |||
AL520811.3 | - | 2, 3 | 130 | |||
BC000098.2 | - | 2, 3 | 156 | 3505930 | ||
BC004274.1 | - | 1, 2 | 81 | 3615320 | ||
BC014885.2 | - | 2, 3 | 125 | 3907411 | ||
BC070169.1 | - | 2, 3 | 156 | 6458566 | ||
BF242439.1 | - | 2, 3 | 243 | |||
BG754026.1 | - | 2, 3 | 204 | |||
BG773256.1 | - | 2, 3 | 232 | |||
BI257891.1 | - | 2, 3 | 187 | |||
BM452844.1 | - | 2, 3 | 110 | |||
BM453663.1 | - | 2, 3 | 99 | |||
CB148943.1 | - | 2, 3 | 183 | |||
DB460745.1 | - | 2, 3 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
