siRNA Details
Gene Symbol: LPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028092.1 | 2 | 203 | |||
NR_028093.1 | 2 | 203 | ||||
GenBank mRNA | U19517.1 | - | 2 | 203 | ||
U19518.1 | - | 2 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
