siRNA Details
Gene Symbol: FASTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258461.1 | NP_001245390.1 | 2 | 553 | ||
NM_006712.4 | NP_006703.1 | 2 | 553 | |||
XM_005249932.1 | XP_005249989.1 | 2 | 745 | |||
XM_005249933.3 | XP_005249990.1 | 2 | 951 | |||
XM_011515761.2 | XP_011514063.1 | 2 | 553 | |||
XM_011515762.2 | XP_011514064.1 | 2 | 553 | |||
XM_017011704.1 | XP_016867193.1 | 2 | 745 | |||
GenBank mRNA | AB209783.1 | - | 2 | 454 | ||
AK023141.1 | - | 2 | 939 | |||
AK223132.1 | - | 2 | 463 | |||
AK292232.1 | - | 2 | 438 | |||
AL713798.1 | - | 1 | 928 | |||
BC000377.2 | - | 2 | 465 | 2820338 | ||
BC011770.2 | - | 2 | 464 | 3831196 | ||
DA253544.1 | - | 2 | 553 | |||
X86779.1 | - | 2 | 457 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023229, AK152369, AK151859, BC013547, AK002552, and others.
Gene Ontology Categories:
