siRNA Details
Gene Symbol: GLT8D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010983.2 | NP_001010983.1 | 4 | 999 | ||
NM_001278280.1 | NP_001265209.1 | 4 | 995 | |||
NM_001278281.1 | NP_001265210.1 | 4 | 743 | |||
NM_018446.3 | NP_060916.1 | 3 | 770 | |||
NM_152932.2 | NP_690909.1 | 3 | 328 | |||
XM_017006857.1 | XP_016862346.1 | 5 | 1158 | |||
XM_017006858.1 | XP_016862347.1 | 4 | 929 | |||
XM_017006859.1 | XP_016862348.1 | 3 | 478 | |||
GenBank mRNA | AF157318.1 | - | 3 | 215 | ||
AF175227.1 | - | 4 | 482 | |||
AF176777.1 | - | 4 | 481 | |||
AJ271710.1 | - | 2 | 97 | |||
AK027517.1 | - | 3 | 236 | |||
AK075031.1 | - | 2 | 292 | |||
AY358579.1 | - | 3 | 249 | |||
BC119670.1 | - | 3 | 214 | 40116196 | ||
BC119671.1 | - | 3 | 214 | 40116197 | ||
BF725274.1 | - | 4 | 416 | |||
CR749821.1 | - | 4 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
