siRNA Details
Gene Symbol: PPP2R1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177562.1 | NP_001171033.1 | 14 | 5392 | ||
NM_001177563.1 | NP_001171034.1 | 13 | 5146 | |||
NM_002716.4 | NP_002707.3 | 15 | 5527 | |||
XM_017017961.1 | XP_016873450.1 | 15 | 2647 | |||
GenBank mRNA | AF163473.1 | - | 15 | 5511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
