siRNA Details
Gene Symbol: TRIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF045239.1 | - | 9 | 1900 | ||
AF220020.1 | - | 9 | 1900 | |||
AF220021.1 | - | 10 | 1867 | |||
AF220022.1 | - | 9 | 1663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
