siRNA Details
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193487.2 | NP_001180416.1 | 5 | 545 | ||
NM_006872.4 | NP_006863.2 | 6 | 647 | |||
GenBank mRNA | AF106857.1 | - | 6 | 567 | ||
AK223350.1 | - | 6 | 571 | |||
AK302220.1 | - | 5 | 528 | |||
BC025991.1 | - | 6 | 596 | 4826221 | ||
BC064585.1 | - | 6 | 551 | |||
BG717289.1 | - | 6 | 622 |
Gene Symbol: STON1-GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198593.1 | NP_001185522.1 | 8 | 2775 | ||
NM_001198594.1 | NP_001185523.1 | 5 | 2570 | |||
NM_172311.2 | NP_758515.1 | 8 | 2778 | |||
GenBank mRNA | AF026169.1 | - | 8 | 2778 | ||
AK309740.1 | - | 8 | 2775 | |||
BC101772.1 | - | 7 | 2691 | 8069278 | ||
BC101774.1 | - | 7 | 2691 | 8069280 | ||
BC143520.1 | - | 5 | 2550 | 9052029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
