siRNA Details
Gene Symbol: PPP2R5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161725.1 | NP_001155197.1 | 16 | 3169 | ||
NM_001161726.1 | NP_001155198.1 | 15 | 3124 | |||
NM_002719.3 | NP_002710.2 | 14 | 3120 | |||
NM_178586.2 | NP_848701.1 | 13 | 3003 | |||
XM_005267819.1 | XP_005267876.1 | 16 | 3230 | |||
XM_005267820.1 | XP_005267877.1 | 16 | 3170 | |||
XM_005267823.1 | XP_005267880.1 | 15 | 3113 | |||
XM_005267824.1 | XP_005267881.1 | 14 | 3040 | |||
XM_005267826.1 | XP_005267883.1 | 14 | 2943 | |||
XM_005267827.1 | XP_005267884.1 | 16 | 3165 | |||
XM_011536914.2 | XP_011535216.1 | 15 | 3034 | |||
XM_011536915.2 | XP_011535217.1 | 15 | 3117 | |||
XM_011536916.2 | XP_011535218.1 | 15 | 4116 | |||
XM_011536918.1 | XP_011535220.1 | 15 | 3271 | |||
XM_011536919.2 | XP_011535221.1 | 15 | 3253 | |||
XM_011536920.1 | XP_011535222.1 | 15 | 3147 | |||
XM_011536921.2 | XP_011535223.1 | 14 | 3036 | |||
XM_017021422.1 | XP_016876911.1 | 13 | 2996 | |||
XM_017021423.1 | XP_016876912.1 | 12 | 2930 | |||
XM_017021424.1 | XP_016876913.1 | 14 | 3030 | |||
GenBank mRNA | AK131391.1 | - | 15 | 3037 | ||
D26445.1 | - | 13 | 2860 | |||
U37352.1 | - | 14 | 3035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
