siRNA Details
Gene Symbol: SOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB020696.1 | - | 1 | 284 | ||
AK022023.1 | - | 1 | 474 | |||
BC113405.1 | - | 1 | 234 | |||
BC113433.1 | - | 1 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
