siRNA Details
Gene Symbol: LIPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 707 | ||
NM_015929.3 | NP_057013.1 | 3 | 509 | |||
NM_145197.2 | NP_660198.1 | 4 | 637 | |||
NM_145198.2 | NP_660199.1 | 4 | 625 | |||
NM_145199.2 | NP_660200.1 | 2 | 383 | |||
NR_037935.1 | 4 | 770 | ||||
NR_037936.1 | 3 | 446 | ||||
GenBank mRNA | AB017566.1 | - | 3 | 509 | ||
BC007001.1 | - | 3 | 429 | 3961227 | ||
BC009772.1 | - | 3 | 450 | 4091817 | ||
BC070145.1 | - | 3 | 487 | 4671058 | ||
BQ422048.1 | - | 3 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
