siRNA Details
Gene Symbol: RGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012720.1 | NP_001012738.1 | 5, 6 | 676 | ||
NM_002921.3 | NP_002912.2 | 5, 6 | 688 | |||
GenBank mRNA | BC011349.1 | - | 5, 6 | 662 | 4157985 | |
BX641033.1 | - | 4, 5 | 1850 | |||
KU178307.1 | - | 5, 6 | 650 | |||
U14910.1 | - | 5, 6 | 676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
