siRNA Details
Gene Symbol: UNC119B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080533.2 | NP_001074002.1 | 5 | 4190 | ||
GenBank mRNA | AK057021.1 | - | 1 | 2324 | ||
AL137327.1 | - | 1 | 1777 | |||
BC004815.1 | - | 1 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
