siRNA Details
Gene Symbol: DDX11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024004.1 | 3 | 2130 | |||
NR_024005.2 | 3 | 1633 | ||||
GenBank mRNA | AK057285.1 | - | 2 | 2133 | ||
AK093685.1 | - | 2 | 3077 | |||
AK123946.1 | - | 3 | 2127 | |||
AM992870.1 | - | 3 | 1633 | |||
BC005070.1 | - | 1 | 705 |
Gene Symbol: DDX11L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070227.1 | - | 1 | 952 |
Gene Symbol: DDX11L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045117.1 | 3 | 1628 | |||
GenBank mRNA | AM992876.1 | - | 3 | 1628 |
Gene Symbol: DDX11L16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110561.1 | 3 | 1616 | |||
GenBank mRNA | AM992882.1 | - | 3 | 1616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
