siRNA Details
Gene Symbol: OSCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282349.1 | NP_001269278.1 | 5 | 1845 | ||
NM_001282350.1 | NP_001269279.1 | 4 | 1812 | |||
NM_130771.4 | NP_570127.3 | 7 | 1343 | |||
NM_133168.4 | NP_573398.2 | 5 | 1298 | |||
NM_133169.4 | NP_573399.2 | 6 | 1331 | |||
NM_206818.2 | NP_996554.2 | 6 | 1857 | |||
GenBank mRNA | AF251702.1 | - | 5 | 1185 | ||
AF391162.1 | - | 5 | 1146 | |||
AF391163.1 | - | 6 | 1151 | |||
AF391164.1 | - | 4 | 1144 | |||
AF474152.1 | - | 4 | 1691 | |||
AF474153.1 | - | 3 | 1620 | |||
AK091111.1 | - | 4 | 1644 | |||
AK130199.1 | - | 5 | 1689 | |||
BC035023.2 | - | 5 | 1845 | 4825982 |
Gene Symbol: KLF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127052.1 | - | 1 | 3204 |
Gene Symbol: LOC105370939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093758.1 | - | 4 | 1419 |
Gene Symbol: PTPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209333.1 | - | 31 | 5129 |
Gene Symbol: SCG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165257.1 | NP_001158729.1 | 11 | 2911 | ||
NM_013243.3 | NP_037375.2 | 12 | 2964 | |||
GenBank mRNA | AF453583.1 | - | 12 | 2969 |
Gene Symbol: VPS26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035260.2 | NP_001030337.1 | 8 | 3292 | ||
NM_001318944.1 | NP_001305873.1 | 10 | 3541 | |||
NM_001318945.1 | NP_001305874.1 | 10 | 3511 | |||
NM_001318946.1 | NP_001305875.1 | 8 | 3359 | |||
NM_004896.4 | NP_004887.2 | 9 | 3435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
