siRNA Details
Gene Symbol: RARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000964.3 | NP_000955.1 | 2 | 621 | ||
NM_001145301.2 | NP_001138773.1 | 2 | 733 | |||
NM_001145302.2 | NP_001138774.1 | 2 | 733 | |||
XM_005257553.1 | XP_005257610.1 | 2 | 525 | |||
XM_005257554.1 | XP_005257611.1 | 2 | 774 | |||
XM_011525095.1 | XP_011523397.1 | 3 | 671 | |||
GenBank mRNA | AK130192.1 | - | 2 | 657 | ||
AK292205.1 | - | 2 | 673 | |||
AK312564.1 | - | 2 | 597 | |||
AL522713.3 | - | 2 | 675 | |||
BC008727.2 | - | 2 | 598 | 3163891 | ||
CR457438.1 | - | 1 | 143 | |||
FJ487576.1 | - | 2 | 669 | |||
HQ692826.1 | - | 1 | 143 | |||
X06538.1 | - | 1 | 369 | |||
X06614.1 | - | 1 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
