siRNA Details
Gene Symbol: CTRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005385.1 | - | 4 | 338 | 3950220 | |
BT007356.1 | - | 3 | 296 | |||
BX497259.1 | - | 4 | 317 | |||
CA946996.1 | - | 3 | 257 | |||
M24400.1 | - | 3 | 312 |
Gene Symbol: CTRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025200.3 | NP_001020371.3 | 4 | 329 | ||
GenBank mRNA | AK124403.1 | - | 2 | 1302 | ||
AK131056.1 | - | 4 | 316 | |||
AK225933.1 | - | 4 | 316 | |||
AK291822.1 | - | 4 | 314 | |||
BC073145.1 | - | 4 | 305 | 5747536 | ||
BP324399.1 | - | 4 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
