siRNA Details
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256799.2 | NP_001243728.1 | 2 | 312 | ||
NM_001289745.1 | NP_001276674.1 | 3 | 370 | |||
NM_001289746.1 | NP_001276675.1 | 2 | 264 | |||
NM_002046.5 | NP_002037.2 | 3 | 278 | |||
GenBank mRNA | AB062273.1 | - | 3 | 142 | ||
AF261085.1 | - | 3 | 193 | |||
AK026525.1 | - | 3 | 215 | |||
AK299448.1 | - | 3 | 163 | |||
AK308198.1 | - | 3 | 166 | |||
AY007133.1 | - | 3 | 142 | |||
AY633612.1 | - | 2 | 114 | |||
BC001601.1 | - | 3 | 144 | 3543589 | ||
BC004109.2 | - | 3 | 142 | 3678259 | ||
BC009081.1 | - | 3 | 151 | 3869809 | ||
BC013310.2 | - | 3 | 150 | 3538383 | ||
BC020308.1 | - | 2 | 128 | |||
BC023632.2 | - | 2 | 114 | 4842418 | ||
BC025925.1 | - | 3 | 147 | 2989348 | ||
BC026907.1 | - | 2 | 128 | 4454083 | ||
BC029340.1 | - | 3 | 136 | |||
BC029618.1 | - | 3 | 144 | 3876535 | ||
BC083511.1 | - | 3 | 142 | 3536969 | ||
BG724119.1 | - | 2 | 295 | |||
BI463134.1 | - | 2 | 294 | |||
BM763361.1 | - | 3 | 255 | |||
BT006893.1 | - | 2 | 90 | |||
BU155402.1 | - | 3 | 315 | |||
CR407671.1 | - | 2 | 90 | |||
DQ403057.1 | - | 1 | 15 | |||
EU668321.1 | - | 3 | 192 | |||
HY000136.1 | - | 2 | 313 | |||
HY004110.1 | - | 2 | 265 | |||
HY022295.1 | - | 2 | 241 | |||
J02642.1 | - | 2 | 91 | |||
M17851.1 | - | 2 | 123 | |||
M33197.1 | - | 3 | 150 | |||
X01677.1 | - | 2 | 150 | |||
X53778.1 | - | 3 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
