siRNA Details
Gene Symbol: RPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032.4 | NP_001023.1 | 2, 3 | 240 | ||
GenBank mRNA | AK026649.1 | - | 1 | 200 | ||
BC032813.1 | - | 1 | 175 | 5217083 | ||
BC035313.1 | - | 175 | 3868721 | |||
BU942244.1 | - | 2, 3 | 243 | |||
L31610.1 | - | 2, 3 | 181 | |||
U14973.1 | - | 2, 3 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
