siRNA Details
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320979.1 | NP_001307908.1 | 1, 2 | 326 | ||
NM_003689.3 | NP_003680.2 | 1, 2 | 326 | |||
GenBank mRNA | AF026947.1 | - | 1, 2 | 294 | ||
AK225267.1 | - | 1, 2 | 297 | |||
AK303131.1 | - | 1, 2 | 262 | |||
AK310609.1 | - | 1, 2 | 283 | |||
BC004111.2 | - | 1, 2 | 253 | 3832140 | ||
BC007352.2 | - | 1, 2 | 301 | |||
BC010852.1 | - | 1, 2 | 302 | 3898226 | ||
BC011586.1 | - | 1, 2 | 253 | 4620484 | ||
BC012171.1 | - | 1, 2 | 301 | 4642811 | ||
BC013996.2 | - | 1, 2 | 257 | 3625337 | ||
BG386786.1 | - | 327 | ||||
BT007347.1 | - | 1, 2 | 217 | |||
BX401279.2 | - | 1, 2 | 274 | |||
EU794591.1 | - | 1, 2 | 326 | |||
Y16675.1 | - | 1, 2 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
