siRNA Details
Gene Symbol: SOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003745.1 | NP_003736.1 | 2 | 232 | ||
GenBank mRNA | AB000676.1 | - | 1 | 36 | ||
AB000734.1 | - | 2 | 232 | |||
AB005043.1 | - | 1 | 131 | |||
AK127621.1 | - | 1 | 800 | |||
U88326.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
