siRNA Details
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002275.2 | NP_001002275.1 | 5, 6 | 882 | ||
NM_001190828.1 | NP_001177757.1 | 4, 5 | 864 | |||
NM_004001.4 | NP_003992.3 | 5, 6 | 885 | |||
XM_017000669.1 | XP_016856158.1 | 6, 7 | 849 | |||
XM_017000670.1 | XP_016856159.1 | 5, 6 | 827 | |||
XM_017000671.1 | XP_016856160.1 | 6, 7 | 849 | |||
GenBank mRNA | AF543826.1 | - | 5, 6 | 838 | ||
BC031992.1 | - | 5, 6 | 795 | 4752609 | ||
BC148273.1 | - | 5, 6 | 780 | 8860219 | ||
M31933.1 | - | 4, 5 | 817 | |||
M31935.1 | - | 5, 6 | 708 | |||
U87560.1 | - | 5, 6 | 758 | |||
U87561.1 | - | 5, 6 | 758 | |||
U87562.1 | - | 5, 6 | 758 | |||
U87563.1 | - | 5, 6 | 758 | |||
U87570.1 | - | 5, 6 | 755 | |||
U87572.1 | - | 5, 6 | 755 | |||
U87573.1 | - | 5, 6 | 758 | |||
U87576.1 | - | 5, 6 | 758 | |||
U87577.1 | - | 5, 6 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
