siRNA Details
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004218.3 | NP_004209.2 | 2 | 209 | ||
GenBank mRNA | AF498947.1 | - | 2 | 113 | ||
AK225036.1 | - | 2 | 127 | |||
AK225642.1 | - | 2 | 129 | |||
AK297498.1 | - | 2 | 137 | |||
AK312994.1 | - | 2 | 163 | |||
BC110081.1 | - | 2 | 115 | 40035112 | ||
BP367041.1 | - | 2 | 209 | |||
BT019535.1 | - | 2 | 113 | |||
BT019536.1 | - | 2 | 113 | |||
BX647356.1 | - | 2211 | ||||
CR536494.1 | - | 2 | 113 | |||
CR541691.1 | - | 2 | 113 | |||
D38516.1 | - | 2 | 122 | |||
EF560724.1 | - | 2 | 192 | |||
X79780.1 | - | 2 | 119 |
