siRNA Details
Gene Symbol: FRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004477.2 | NP_004468.1 | 5, 6 | 620 | ||
GenBank mRNA | AK291890.1 | - | 5, 6 | 594 | ||
BC053997.1 | - | 5, 6 | 577 | 5555121 | ||
L76159.1 | - | 5, 6 | 620 |
Gene Symbol: FRG1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003579.1 | 4, 5 | 525 | |||
GenBank mRNA | BC062743.1 | - | 5, 6 | 744 | ||
BC095491.1 | - | 4, 5 | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
