siRNA Details
Gene Symbol: KRTAP5-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005553.3 | NP_005544.4 | 1 | 498 | ||
GenBank mRNA | AB126078.1 | - | 1 | 294 | ||
BC069531.1 | - | 1 | 260 | 7262098 | ||
BC101744.1 | - | 1 | 402 | 8069250 | ||
X63755.1 | - | 1 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
