siRNA Details
Gene Symbol: SPRR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003125.2 | NP_003116.2 | 2 | 336 | ||
GenBank mRNA | BC056240.1 | - | 2 | 340 | ||
M19888.1 | - | 2 | 340 | |||
M21300.1 | - | 2 | 336 | |||
M21301.1 | - | 2 | 336 |
Gene Symbol: SPRR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199828.1 | NP_001186757.1 | 2 | 343 | ||
NM_005987.3 | NP_005978.2 | 2 | 329 | |||
GenBank mRNA | BC105081.1 | - | 2 | 343 | 8144084 | |
BC105083.1 | - | 2 | 329 | 8144086 | ||
BC143782.1 | - | 2 | 343 | 9052296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
