siRNA Details
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320979.1 | NP_001307908.1 | 1, 2 | 328 | ||
NM_003689.3 | NP_003680.2 | 1, 2 | 328 | |||
GenBank mRNA | AF026947.1 | - | 1, 2 | 296 | ||
AK225267.1 | - | 1, 2 | 299 | |||
AK303131.1 | - | 1, 2 | 264 | |||
AK310609.1 | - | 1, 2 | 285 | |||
BC004111.2 | - | 1, 2 | 255 | 3832140 | ||
BC007352.2 | - | 1, 2 | 303 | |||
BC010852.1 | - | 1, 2 | 304 | 3898226 | ||
BC011586.1 | - | 1, 2 | 255 | 4620484 | ||
BC012171.1 | - | 1, 2 | 303 | 4642811 | ||
BC013996.2 | - | 1, 2 | 259 | 3625337 | ||
BG386786.1 | - | 329 | ||||
BT007347.1 | - | 1, 2 | 219 | |||
BX401279.2 | - | 1, 2 | 276 | |||
EU794591.1 | - | 1, 2 | 328 | |||
Y16675.1 | - | 1, 2 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
