siRNA Details
Gene Symbol: CLCNKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042704.1 | NP_001036169.1 | 3 | 279 | ||
NM_001257139.1 | NP_001244068.1 | 3 | 279 | |||
NM_004070.3 | NP_004061.3 | 3 | 279 | |||
GenBank mRNA | AK225550.1 | - | 3 | 234 | ||
AK290814.1 | - | 3 | 234 | |||
AK298285.1 | - | 3 | 258 | |||
BC035373.1 | - | 7 | 858 | |||
BC053869.1 | - | 4 | 327 | 5187202 | ||
Z30643.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
