siRNA Details
Gene Symbol: ZNF443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005815.4 | NP_005806.2 | 4 | 1407 | ||
XM_011527622.2 | XP_011525924.1 | 4 | 1934 | |||
XM_011527623.2 | XP_011525925.2 | 4 | 2475 | |||
XM_017026139.1 | XP_016881628.1 | 4 | 2475 | |||
XM_017026140.1 | XP_016881629.1 | 3 | 2978 | |||
GenBank mRNA | AB011414.1 | - | 4 | 1313 | ||
AK023025.1 | - | 4 | 1289 | |||
AK299532.1 | - | 4 | 1244 | |||
BC032753.1 | - | 4 | 1311 | 5583971 |
